SY3H3D6811E3 Synthonix Corporation

5-Nitrobenzo[d]thiazol-2-amine

  • MDL number No MDL found
  • Cas Number : 73458-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/44886.html

Safety Information

MSDS https://synthonix.com/44886.html
Availability details for
Quantity
  • Availability
  • Add to Cart