SY3H3D6811E9 Synthonix Corporation

1-(3-Nitropyridin-4-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 161871-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45246.html

Safety Information

MSDS https://synthonix.com/45246.html
Availability details for
Quantity
  • Availability
  • Add to Cart