SY3H3D6811EE Synthonix Corporation

5-Nitro-2-(propylthio)pyrimidine-4,6-diol

  • MDL number MFCD26385870
  • Cas Number : 145783-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46357.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46357.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart