SY3H3D6811F7 Synthonix Corporation

6-Nitroquinoxaline-2,3-dione

  • Cas Number : 2379-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47267.html

Safety Information

MSDS https://synthonix.com/47267.html
Availability details for
Quantity
  • Availability
  • Add to Cart