SY3H3D681200 Synthonix Corporation

4-Nitro-2-(trifluoromethyl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 50551-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47421.html

Safety Information

MSDS https://synthonix.com/47421.html
Availability details for
Quantity
  • Availability
  • Add to Cart