SY3H3D68120B Synthonix Corporation

6-Nitroquinoxalin-2-one

  • MDL number No MDL found
  • Cas Number : 25652-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47645.html

Safety Information

MSDS https://synthonix.com/47645.html
Availability details for
Quantity
  • Availability
  • Add to Cart