SY3H3D68120D Synthonix Corporation

7-Nitro-2(1H)-quinoxalinone

  • MDL number No MDL found
  • Cas Number : 89898-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47647.html

Safety Information

MSDS https://synthonix.com/47647.html
Availability details for
Quantity
  • Availability
  • Add to Cart