SY3H3D681211 Synthonix Corporation

6-Nitroquinazoline-2,4-diol

  • MDL number No MDL found
  • Cas Number : 32618-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47651.html

Safety Information

MSDS https://synthonix.com/47651.html
Availability details for
Quantity
  • Availability
  • Add to Cart