SY3H3D681213 Synthonix Corporation

5-Nitrobenzo[b]thiophene

  • MDL number No MDL found
  • Cas Number : 4965-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/47720.html

Safety Information

MSDS https://synthonix.com/47720.html
Availability details for
Quantity
  • Availability
  • Add to Cart