SY3H3D68121B Synthonix Corporation

1,5-Naphthyridine-4,8-diol

  • Cas Number : 28312-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47874.html

Safety Information

MSDS https://synthonix.com/47874.html
Availability details for
Quantity
  • Availability
  • Add to Cart