SY3H3D68160B Synthonix Corporation

Pyrrolidin-3-ylmethanol hydrochloride hydrate

  • MDL number No MDL found
  • Cas Number : 1452519-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41328.html

Safety Information

MSDS https://synthonix.com/41328.html
Availability details for
Quantity
  • Availability
  • Add to Cart