SY3H3D68160F Synthonix Corporation

3-Phenoxypiperidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1171992-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55490.html

Safety Information

MSDS https://synthonix.com/55490.html
Availability details for
Quantity
  • Availability
  • Add to Cart