SY3H3D681612 Synthonix Corporation

5-Nitro-2,3-dihydrobenzofuran

  • MDL number No MDL found
  • Cas Number : 17403-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48504.html

Safety Information

MSDS https://synthonix.com/48504.html
Availability details for
Quantity
  • Availability
  • Add to Cart