SY3H3D681618 Synthonix Corporation

6-Nitro-1,2,3,4-tetrahydroquinoxaline

  • MDL number No MDL found
  • Cas Number : 41959-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49217.html

Safety Information

MSDS https://synthonix.com/49217.html
Availability details for
Quantity
  • Availability
  • Add to Cart