SY3H3D68161B Synthonix Corporation

(R)-1-(3-Nitrophenyl)ethanol

  • MDL number No MDL found
  • Cas Number : 76116-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49315.html

Safety Information

MSDS https://synthonix.com/49315.html
Availability details for
Quantity
  • Availability
  • Add to Cart