SY3H3D68161C Synthonix Corporation

(R)-1-(4-Nitrophenyl)ethanol

  • MDL number No MDL found
  • Cas Number : 58287-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49316.html

Safety Information

MSDS https://synthonix.com/49316.html
Availability details for
Quantity
  • Availability
  • Add to Cart