SY3H3D681621 Synthonix Corporation

7-Nitro-1,2,3,4-tetrahydroisoquinoline

  • MDL number No MDL found
  • Cas Number : 42923-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49736.html

Safety Information

MSDS https://synthonix.com/49736.html
Availability details for
Quantity
  • Availability
  • Add to Cart