SY3H3D681623 Synthonix Corporation

N-(4-Nitrophenyl)propionamide

  • MDL number No MDL found
  • Cas Number : 4850-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49739.html

Safety Information

MSDS https://synthonix.com/49739.html
Availability details for
Quantity
  • Availability
  • Add to Cart