SY3H3D681626 Synthonix Corporation

(E)-3-(5-Nitrocyclohex-1-en-1-yl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 899809-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50109.html

Safety Information

MSDS https://synthonix.com/50109.html
Availability details for
Quantity
  • Availability
  • Add to Cart