SY3H3D681628 Synthonix Corporation

Non-8-yn-1-ol

  • MDL number No MDL found
  • Cas Number : 10160-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50499.html

Safety Information

MSDS https://synthonix.com/50499.html
Availability details for
Quantity
  • Availability
  • Add to Cart