SY3H3D681631 Synthonix Corporation

8-Nitroquinolin-6-ol

  • MDL number No MDL found
  • Cas Number : 5437-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50821.html

Safety Information

MSDS https://synthonix.com/50821.html
Availability details for
Quantity
  • Availability
  • Add to Cart