SY3H3D681632 Synthonix Corporation

4-(4-Nitrophenyl)oxazole

  • MDL number No MDL found
  • Cas Number : 13382-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50822.html

Safety Information

MSDS https://synthonix.com/50822.html
Availability details for
Quantity
  • Availability
  • Add to Cart