SY3H3D681634 Synthonix Corporation

7-Nitroisoquinolin-1(2H)-one

  • MDL number No MDL found
  • Cas Number : 20141-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50825.html

Safety Information

MSDS https://synthonix.com/50825.html
Availability details for
Quantity
  • Availability
  • Add to Cart