SY3H3D68163A Synthonix Corporation

4-Nitro-1-phenyl-1H-imidazole

  • MDL number No MDL found
  • Cas Number : 41384-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/51142.html

Safety Information

MSDS https://synthonix.com/51142.html
Availability details for
Quantity
  • Availability
  • Add to Cart