SY3H3D68163C Synthonix Corporation

3-(3-Nitrophenyl)isoxazol-5-amine

  • MDL number No MDL found
  • Cas Number : 887591-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51150.html

Safety Information

MSDS https://synthonix.com/51150.html
Availability details for
Quantity
  • Availability
  • Add to Cart