SY3H3D68163D Synthonix Corporation

5-Nitro-1H-inden-2(3H)-one

  • Cas Number : 116530-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51198.html

Safety Information

MSDS https://synthonix.com/51198.html
Availability details for
Quantity
  • Availability
  • Add to Cart