SY3H3D68163F Synthonix Corporation

7-Nitro-1,4-benzodioxane-6-carboxylic Acid

  • MDL number No MDL found
  • Cas Number : 57672-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51235.html

Safety Information

MSDS https://synthonix.com/51235.html
Availability details for
Quantity
  • Availability
  • Add to Cart