SY3H3D681642 Synthonix Corporation

2-(4-Nitrophenyl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 19910-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51828.html

Safety Information

MSDS https://synthonix.com/51828.html
Availability details for
Quantity
  • Availability
  • Add to Cart