SY3H3D681646 Synthonix Corporation

3-(3-Nitrophenoxy)propionic acid

  • Cas Number : 91004-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51850.html

Safety Information

MSDS https://synthonix.com/51850.html
Availability details for
Quantity
  • Availability
  • Add to Cart