SY3H3D681647 Synthonix Corporation

3-(4-Nitrophenoxy)propanoic acid

  • MDL number No MDL found
  • Cas Number : 10572-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51851.html

Safety Information

MSDS https://synthonix.com/51851.html
Availability details for
Quantity
  • Availability
  • Add to Cart