SY3H3D681648 Synthonix Corporation

1-(6-Nitrobenzo[d][1,3]dioxol-5-yl)ethanol

  • Cas Number : 159873-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51861.html

Safety Information

MSDS https://synthonix.com/51861.html
Availability details for
Quantity
  • Availability
  • Add to Cart