SY3H3D68164A Synthonix Corporation

1,6-Naphthyridine-2-carboximidamide hydrochloride

  • MDL number No MDL found
  • Cas Number : 1179360-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52222.html

Safety Information

MSDS https://synthonix.com/52222.html
Availability details for
Quantity
  • Availability
  • Add to Cart