SY3H3D68164E Synthonix Corporation

2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol

  • MDL number MFCD00239474
  • Cas Number : 84041-77-0
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Light-brown to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53132.html

Safety Information

signalword Warning
pcodes P261,P280,P305,P351,P338
MSDS https://synthonix.com/53132.html
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart