SY3H3D68164F Synthonix Corporation

6-Nitro-4-(trifluoromethyl)quinolin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 328956-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53390.html

Safety Information

MSDS https://synthonix.com/53390.html
Availability details for
Quantity
  • Availability
  • Add to Cart