SY3H3D681650 Synthonix Corporation

5-Nitroquinoline-8-carbonitrile

  • MDL number No MDL found
  • Cas Number : 205046-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53396.html

Safety Information

MSDS https://synthonix.com/53396.html
Availability details for
Quantity
  • Availability
  • Add to Cart