SY3H3D681651 Synthonix Corporation

5-(3-Nitrophenyl)isoxazole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 199601-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53435.html

Safety Information

MSDS https://synthonix.com/53435.html
Availability details for
Quantity
  • Availability
  • Add to Cart