SY3H3D681654 Synthonix Corporation

4-Nitro-1-phenyl-1H-pyrazole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 701917-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53547.html

Safety Information

MSDS https://synthonix.com/53547.html
Availability details for
Quantity
  • Availability
  • Add to Cart