SY3H3D681658 Synthonix Corporation

4-(3-Nitrophenyl)pyrimidin-2-amine

  • MDL number No MDL found
  • Cas Number : 240136-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53695.html

Safety Information

MSDS https://synthonix.com/53695.html
Availability details for
Quantity
  • Availability
  • Add to Cart