SY3H3D6816CC Synthonix Corporation

4-(4-Nitrophenyl)but-3-en-2-one

  • MDL number MFCD00661313
  • Cas Number : 3490-37-7
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53956.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/53956.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart