SY3H3D6816CE Synthonix Corporation

5-Nitro-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 51114-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53964.html

Safety Information

MSDS https://synthonix.com/53964.html
Availability details for
Quantity
  • Availability
  • Add to Cart