SY3H3D6816EA Synthonix Corporation

4-Nitrophenyl 5-chlorothiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1450877-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55171.html

Safety Information

MSDS https://synthonix.com/55171.html
Availability details for
Quantity
  • Availability
  • Add to Cart