SY3H3D6816EC Synthonix Corporation

4-Nitro-1-naphthoic acid

  • MDL number No MDL found
  • Cas Number : 1975-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55196.html

Safety Information

MSDS https://synthonix.com/55196.html
Availability details for
Quantity
  • Availability
  • Add to Cart