SY3H3D6816FA Synthonix Corporation

4-((2-Nitrophenyl)amino)phenol

  • MDL number No MDL found
  • Cas Number : 54381-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55568.html

Safety Information

MSDS https://synthonix.com/55568.html
Availability details for
Quantity
  • Availability
  • Add to Cart