SY3H3D681706 Synthonix Corporation

5-Nitro-2-(piperidin-1-yl)benzonitrile

  • MDL number No MDL found
  • Cas Number : 32188-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/55798.html

Safety Information

MSDS https://synthonix.com/55798.html
Availability details for
Quantity
  • Availability
  • Add to Cart