SY3H3D68170C Synthonix Corporation

1-(4-Nitrophenyl)piperidin-4-yl methanesulfonate

  • MDL number No MDL found
  • Cas Number : 303977-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56044.html

Safety Information

MSDS https://synthonix.com/56044.html
Availability details for
Quantity
  • Availability
  • Add to Cart