SY3H3D681716 Synthonix Corporation

3-(Naphthalen-1-yl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 13026-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56678.html

Safety Information

MSDS https://synthonix.com/56678.html
Availability details for
Quantity
  • Availability
  • Add to Cart