SY3H3D681718 Synthonix Corporation

Naphtho[1,2-b]thiophen-2-ylmethanol

  • MDL number No MDL found
  • Cas Number : 35813-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56691.html

Safety Information

MSDS https://synthonix.com/56691.html
Availability details for
Quantity
  • Availability
  • Add to Cart