SY3H3D68171A Synthonix Corporation

3-(4-Nitro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

  • MDL number No MDL found
  • Cas Number : 827026-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56705.html

Safety Information

MSDS https://synthonix.com/56705.html
Availability details for
Quantity
  • Availability
  • Add to Cart