SY3H3D681720 Synthonix Corporation

2-(Naphthalen-1-yl)propan-2-ol

  • MDL number No MDL found
  • Cas Number : 6301-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56879.html

Safety Information

MSDS https://synthonix.com/56879.html
Availability details for
Quantity
  • Availability
  • Add to Cart