SY3H3D681721 Synthonix Corporation

1-(3-(4-Nitrophenoxy)propyl)pyrrolidine

  • MDL number No MDL found
  • Cas Number : 92033-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/57080.html

Safety Information

MSDS https://synthonix.com/57080.html
Availability details for
Quantity
  • Availability
  • Add to Cart